T8785
Thyrotropic hormone from porcine pituitary
5 IU/mg
Synonym(s):
TSH, Thyroid stimulating hormone
About This Item
Recommended Products
biological source
Porcine pituitary glands
Quality Level
form
powder
specific activity
5 IU/mg
packaging
vial of 10 IU
technique(s)
cell culture | mammalian: suitable
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... TSHR(7253)
pig ... TSH-BETA(397658)
Application
Biochem/physiol Actions
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service