Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T8537

Sigma-Aldrich

Anti-Tissue Inhibitor of Metalloproteinase-4, First Loop antibody produced in rabbit

~1 mg/mL, buffered aqueous glycerol solution, affinity isolated antibody

Synonym(s):

Anti-TIMP-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

SMILES string

CCCCCCCCCSCCCCCCCC(=O)OC

Gene Information

human ... TIMP4(7079)

Specificity

Reacts with reduced and non-reduced TIMP-4. By immunoblotting against the reduced protein, the antibody reacts with bands at approximately 23 kDa and 29 kDa (unglycosylated and glycosylated, respectively).

Immunogen

synthetic peptide corresponding to the first loop of human TIMP-4.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service