Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T8156

Sigma-Aldrich

Adrenorphin

≥95% (HPLC)

Synonym(s):

Metorphamide, Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H68N14O10S
CAS Number:
Molecular Weight:
985.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26
assay:
≥95% (HPLC)

assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C(C)C)C(O)=O

InChI

1S/C44H68N14O10S/c1-25(2)36(42(67)68)58-40(65)31(12-8-19-51-44(48)49)56-38(63)30(11-7-18-50-43(46)47)55-39(64)32(17-20-69-3)57-41(66)33(22-26-9-5-4-6-10-26)54-35(61)24-52-34(60)23-53-37(62)29(45)21-27-13-15-28(59)16-14-27/h4-6,9-10,13-16,25,29-33,36,59H,7-8,11-12,17-24,45H2,1-3H3,(H,52,60)(H,53,62)(H,54,61)(H,55,64)(H,56,63)(H,57,66)(H,58,65)(H,67,68)(H4,46,47,50)(H4,48,49,51)/t29-,30-,31-,32-,33-,36-/m0/s1

InChI key

ZBRXWGQCHSFAHM-MWSMAVIUSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S J Bunn et al.
Biochemical pharmacology, 41(5), 715-722 (1991-03-01)
The ability of a number of opioid agonists and antagonists to affect nicotine-induced 45Ca2(+)-uptake into cultured bovine adrenal medullary cells has been investigated. High (10 microM) concentrations of the opioid agonist bremazocine produced a significant inhibition of nicotine-induced 45Ca2(+)-uptake throughout
Vadivel Ganapathy et al.
The AAPS journal, 7(4), E852-E856 (2006-04-06)
Transmembrane transport of endogenous as well as synthetic opioid peptides is a critical determinant of pharmacokinetics and biologic efficacy of these peptides. This transport process influences the distribution of opioid peptides across the blood-brain barrier and their elimination from the
T Yanase et al.
The Journal of clinical endocrinology and metabolism, 64(4), 692-697 (1987-04-01)
We studied the secretion and tissue contents of adrenorphin in human pheochromocytomas. In 17 human pheochromocytomas from 11 patients, we found a remarkably wide distribution in immunoreactive adrenorphin levels (3-7771 pg/mg tissue). Adrenomedullary pheochromocytomas contained a significantly larger amount of
P D Marley et al.
Brain research, 363(1), 10-17 (1986-01-15)
Opioid peptides are found in high concentrations in the adrenal medulla. Recently, a novel opioid octapeptide, metorphamide, possessing an amidated C-terminal, was characterized and also found to be present in adrenal tissue. We have studied the ability of this novel
Fuyu Guan et al.
Analytica chimica acta, 985, 79-90 (2017-09-03)
The ability to analyze biological samples for multitudinous exogenous peptides with a single analytical method is desired for doping control in horse racing. The key to achieving this goal is the capability of extracting all target peptides from the sample

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service