T7817
Standard PCR microtubes
PCR tube (Domed cap), capacity 0.2 mL
About This Item
Recommended Products
material
V-bottom bottom
colorless tube
domed cap
natural
polypropylene
polypropylene tube
straight neck
feature
RNase and DNase free
neck thread push fit
packaging
pack of 1000 ea
manufacturer/tradename
Thermo Scientific AB-0337
technique(s)
PCR: suitable
capacity
0.2 mL
General description
Certified free from DNase, RNase and human genomic DNA contamination.
Suitable for use in all 96 well ′v′ bottomed thermal cyclers
Integral snap-shut domed cap
Choose from one of the most recent versions:
Certificates of Analysis (COA)