T7580
TN-16
≥98% (HPLC)
Synonym(s):
3-[1-(Phenylamino)ethylidene]-5-(phenylmethyl)-2,4-pyrrolidinedione, NSC 239274
Select a Size
Select a Size
About This Item
Recommended Products
Quality Level
assay
≥98% (HPLC)
form
powder
color
white to tan
solubility
DMSO: ≥17 mg/mL
storage temp.
2-8°C
SMILES string
C\C(Nc1ccccc1)=C2\C(=O)NC(Cc3ccccc3)C2=O
InChI
1S/C19H18N2O2/c1-13(20-15-10-6-3-7-11-15)17-18(22)16(21-19(17)23)12-14-8-4-2-5-9-14/h2-11,16,20H,12H2,1H3,(H,21,23)/b17-13-
InChI key
KWSXUGYAGMSUTN-LGMDPLHJSA-N
Application
Biochem/physiol Actions
signalword
Warning
hcodes
Hazard Classifications
Aquatic Acute 1 - Skin Sens. 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service