Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7444

Sigma-Aldrich

Anti-phospho-Tau (pSer404) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

phosphorylation (pSer404)

Gene Information

human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)

General description

Tau is a microtubule-associated phosphoprotein that is present in the axons of nerve cells. It stabilizes microtubules and mediates the polymerization of tubulin. Phosphorylation of Tau regulates its cellular functions. Hyperphosphorylation of Tau has been linked to neurodegenerative disorders such as Alzheimer′s disease . Anti-phospho-Tau (pSer404) antibody specifically reacts with Tau (pSer404) (45-68 kDa) in humans, mice and rats.

Immunogen

chemically synthesized phosphopeptide derived from the region of human tau that contains serine404.

Application

Anti-phospho-Tau (pSer404) antibody may be used in western blot (1:1000), immunoblot (1:1000) and immunohistochemistry.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 50% glycerol, 1.0 mg/ml BSA (IgG and protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Haque et al.
Brain research, 838(1-2), 69-77 (1999-08-14)
Tau, one of the best characterized microtubule-associated proteins (MAPs), is a phosphoprotein, the biological activity of which is regulated by its degree of phosphorylation. The objective of the present study was to evaluate the regulation, phosphorylation and the biological activity
Océane Guillemaud et al.
Neurobiology of aging, 90, 135-146 (2020-03-17)
In Alzheimer disease (AD), astrocytes undergo complex changes and become reactive. The consequences of this reaction are still unclear. To evaluate the net impact of reactive astrocytes in AD, we developed viral vectors targeting astrocytes that either activate or inhibit
S M Jenkins et al.
The Biochemical journal, 345 Pt 2, 263-270 (2000-01-06)
Tau is a microtubule-associated protein that is functionally modulated by phosphorylation and hyperphosphorylated in several neurodegenerative diseases. Because phosphorylation regulates both normal and pathological tau functioning, it is of great interest to identify the signalling pathways and enzymes capable of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service