direct immunofluorescence: 1:16 immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:160
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Application
Conjugation of TRITC to Guinea Pig IgG (whole molecule) antibody enables its use in immunofluorescent techniques.
Biochem/physiol Actions
IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Regulatory T-cells (Tregs) play a pivotal role in maintaining peripheral tolerance. Increasing Treg numbers/functions has been shown to ameliorate autoimmune diseases. However, common Treg expansion approaches use T-Cell Receptor (TCR)-mediated stimulation which also causes proliferation of effector T-cells (Teff). To
Pancreatic β-cell dysfunction is a common feature of type 2 diabetes. Earlier, we had cloned IG20 cDNA from a human insulinoma and had shown that IG20/MADD can encode six different splice isoforms that are differentially expressed and have unique functions
We present a simple method to pattern capture antibodies (cAbs) on poly(dimethylsiloxane) (PDMS), with high accuracy and in a manner compatible with mass fabrication for use with capillary systems (CSs), using stencils microfabricated in Si. Capture antibodies are patterned as
We have examined factors affecting the in vitro differentiation of Pdx1(GFP/w) ESCs to pancreatic endocrine cells. Inclusion of Bone Morphogenetic Protein 4 (BMP4) during the first four days of differentiation followed by a 24-hour pulse of retinoic acid (RA) induced
British journal of haematology, 118(1), 108-116 (2002-07-09)
Graft-versus-host disease (GvHD) is a multistep immune process involving lymphocyte activation, proliferation and target cell killing. We have devised a novel method for the selective depletion of alloreactive cells from haematopoietic stem cell grafts which retains a pool of immunocompetent
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.