Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T7066

Sigma-Aldrich

Anti-Transglutaminase Type II antibody produced in goat

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen ~78 kDa

species reactivity

guinea pig, mouse

technique(s)

immunoprecipitation (IP): 6 μL using 0.5-1 mg of murine liver or spleen tissue extract
western blot: 1:650 using murine liver, testis, thymus tissue extracts

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Tgm2(21817)

Immunogen

guinea pig liver type II transglutaminase

Physical form

Whole antiserum containing 0.05% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chien-Wei Liao et al.
BMC infectious diseases, 8, 84-84 (2008-06-25)
Because the outcomes and sequelae after different types of brain injury (BI) are variable and difficult to predict, investigations on whether enhanced expressions of BI-associated biomarkers (BIABs), including transforming growth factor beta1 (TGF-beta1), S100B, glial fibrillary acidic protein (GFAP), neurofilament
Chia-Mei Chou et al.
Parasites & vectors, 10(1), 620-620 (2017-12-24)
Toxocariasis is a worldwide zoonotic parasitic disease mainly caused by Toxocara canis. Humans can be infected by accidental ingestion of T. canis embryonated ovum-contaminated food, water, or encapsulated larvae in paratenic hosts' viscera or meat. Since humans and mice are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service