Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6570

Sigma-Aldrich

Tissue protein, cytoplasmic

from human skeletal muscle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human skeletal muscle

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Highly purified cytoplasmic protein from whole tissue homogenates. Useful in electrophoresis, Western blotting, immunoassay, and other applications requiring source-specific, non-denatured protein.

Physical form

3 mg protein (approx. 10 mg protein/mL) in a solution containing 20 mM HEPES, pH 7.9, 1.5 mM MgCl2, 10 mM KCl, 1 mM EDTA, 200 mM sucrose, 10% glycerol, and a cocktail of protease inhibitor.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service