Key Documents
T6531
Sigma-Aldrich
Gel casting tray for horizontal submarine units
size 20 cm × 20 cm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
size
20 cm × 20 cm
Features and Benefits
Construction of 1/4 in. thick UV-transparent acrylic prevents distortion during gel formation and permits use on a UV transilluminator. Grooves provide for uniform placement of combs near either end or toward the center of the gel. Wells in base form agarose "anchors," preventing gel from floating. Extra trays permit pouring one gel while running another.
Choose from one of the most recent versions:
Certificates of Analysis (COA)