biological source
rat spleen
form
liquid
shipped in
dry ice
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
Application
Highly purified cell membrane protein from whole tissue homogenates. Useful in electrophoresis, Western blotting, immunoassay, and other applications requiring source-specific, non-denatured protein.
Physical form
0.5 mg protein (approx. 3 mg protein/mL)
Supplied in a solution containing 20 mM HEPES, 1.5 mM MgCl2, 10 mM KCl, 1 mM EDTA, 2% glycerol, 0.5% sodium deoxycholate, 1% NP-40, pH 7.9, and a cocktail of protease inhibitors.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service