Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

T6259

Sigma-Aldrich

DL-4-Thiaisoleucine

~98% (TLC)

Synonym(s):

2-Amino-3-(methylthio)butyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO2S
CAS Number:
Molecular Weight:
149.21
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

DL-4-Thiaisoleucine,

assay

~98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CSC(C)C(N)C(O)=O

InChI

1S/C5H11NO2S/c1-3(9-2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)

InChI key

PWDYHRMBGRYCAP-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva S Istvan et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(4), 1627-1632 (2011-01-06)
Intraerythrocytic malaria parasites can obtain nearly their entire amino acid requirement by degrading host cell hemoglobin. The sole exception is isoleucine, which is not present in adult human hemoglobin and must be obtained exogenously. We evaluated two compounds for their
Ian J Clifton et al.
Chembiochem : a European journal of chemical biology, 12(12), 1881-1885 (2011-06-17)
Isopenicillin N synthase (IPNS) catalyses the synthesis of isopenicillin N (IPN), the biosynthetic precursor to penicillin and cephalosporin antibiotics. IPNS is a non-heme iron(II) oxidase that mediates the oxidative cyclisation of the tripeptide δ-L-α-aminoadipoyl-L-cysteinyl-D-valine (ACV) to IPN with a concomitant
M Green
The Journal of biological chemistry, 257(15), 9039-9042 (1982-08-10)
The incorporation of the leucine analog, beta-hydroxyleucine, or the isoleucine analog, 4-thiaisoleucine, into the MPOC-46B kappa chain interfered with the processing of the oligosaccharide moiety of newly synthesized light chains. Analog-containing chains were secreted with endo-beta-N-acetylglucosaminidase H-sensitive oligosaccharides instead of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service