Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6075

Sigma-Aldrich

Anti-Tachykinin Receptor 2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunocytochemistry: 5 μg/mL using human hyperplastic smooth muscle tissue
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5 μg/mL using human hyperplastic smooth muscle tissue

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TACR2(6865)
mouse ... Tacr2(21337)
rat ... Tacr2(25007)

Immunogen

synthetic peptide corresponding to the C-terminal domain of human tachykinin receptor 2, conjugated to KLH. The immunizing peptide has 85% homology with the rat and mouse gene.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service