Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T6001

Sigma-Aldrich

2′,3′,5′-Tri-O-acetyluridine

≥98%

Synonym(s):

Uridine 2′,3′,5′-triacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N2O9
CAS Number:
Molecular Weight:
370.31
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

biological source

synthetic (organic)

assay

≥98%

form

powder

solubility

chloroform: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(O[C@H]1[C@H](N2C=CC(NC2=O)=O)O[C@H](COC(C)=O)[C@H]1OC(C)=O)=O

InChI

1S/C15H18N2O9/c1-7(18)23-6-10-12(24-8(2)19)13(25-9(3)20)14(26-10)17-5-4-11(21)16-15(17)22/h4-5,10,12-14H,6H2,1-3H3,(H,16,21,22)/t10?,12-,13-,14-/m1/s1

InChI key

AUFUWRKPQLGTGF-FYFXECOGSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra Krstulja et al.
Analytical and bioanalytical chemistry, 406(25), 6275-6284 (2014-08-01)
In this paper, we describe the synthesis and evaluation of molecularly imprinted polymers (MIPs), prepared using 2',3',5'-tri-O-acyluridines as 'dummy' templates, for the selective recognition of uridine nucleosides. The MIPs were synthesised using a non-covalent approach with 2,6-bis-acrylamidopyridine (BAAPy) acting as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service