Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5943

Sigma-Aldrich

Tyrphostin AG 1007

≥98%, solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2O2S
CAS Number:
Molecular Weight:
310.37
MDL number:
UNSPSC Code:
51112000
NACRES:
NA.77

Quality Level

assay

≥98%

form

solid

color

yellow to green

solubility

DMSO: soluble
acetone: soluble

storage temp.

−20°C

Biochem/physiol Actions

Tyrphostin AG 1007 is known to function as a receptor tyrosine kinase inhibitor. It has been reported that replacement of oxygen in the carbonyl group of AG-1007 significantly increased the inhibition of tyrosine autophosphorylation of IGF-1R1.

Preparation Note

Tyrphostin AG 1007 is soluble in DMSO and acetone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Blum et al.
Biochemistry, 39(51), 15705-15712 (2000-12-22)
IGF-1 and its receptor play a pivotal role in many cancers, and therefore, IGF-1R is an attractive target for the design of inhibitors. In this communication, we report on a number of lead compounds for inhibitors of the isolated IGF-1R

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service