T5819
TRAIL Receptor 2/Fc Chimera from mouse
>95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder
Synonym(s):
DR5, MK, TRICK2
About This Item
recombinant
expressed in baculovirus infected Sf21 cells
assay
>95% (SDS-PAGE)
form
lyophilized powder
mol wt
~49 kDa by SDS-PAGE (reducing)
calculated mol wt ~40 kDa
impurities
endotoxin, tested
UniProt accession no.
storage temp.
−20°C
Gene Information
mouse ... Tnfrsf10b(21933)
Application
Biochem/physiol Actions
Other Notes
Physical form
Analysis Note
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service