Key Documents
T5541
Supelco
Glass tubes for tube gels
I.D. × L 1.5 mm × 7.5 cm
About This Item
UNSPSC Code:
12352200
I.D. × L
1.5 mm × 7.5 cm
Quality Level
Features and Benefits
Although the tubes listed below have different inside diameters, they all have the same 3.0 mm outside diameter. This makes the tubes less fragile and easier to handle.
Other Notes
The 7.5 cm tubes fit the mini electrophoresis unit, and the 13.5 cm tubes fit the standard-size unit.
Choose from one of the most recent versions:
Certificates of Analysis (COA)