Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5129

Sigma-Aldrich

Tyr-Ala

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O4
CAS Number:
Molecular Weight:
252.27
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

assay

≥98% (TLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

OC([C@H](C)NC([C@@H](N)CC1=CC=C(O)C=C1)=O)=O

InChI

1S/C12H16N2O4/c1-7(12(17)18)14-11(16)10(13)6-8-2-4-9(15)5-3-8/h2-5,7,10,15H,6,13H2,1H3,(H,14,16)(H,17,18)

InChI key

NLKUJNGEGZDXGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Purdie et al.
Chirality, 11(7), 546-553 (1999-07-28)
Complexation with Cu(II) ion in strong aqueous base, combined with visible range circular dichroism detection, were used to quantitatively differentiate among the L-enantiomers of the GG, GA, GY, AG, AA, AY, YG, YA, and YY dipeptides and the D-enantiomer of
Guang Huang et al.
Environmental science & technology, 53(7), 3672-3680 (2019-02-27)
Dipeptides are widely present in surface water and serve as precursors to form disinfection byproducts (DBPs) during disinfection (e.g., chloramination). Bromide (Br-) and iodide (I-) are common in many source waters, enhancing Br- and I-DBP formation. Recently Cl-, I-, and
Spectroscopic and structural elucidation of L-tyrosine-containing dipeptides valyl-tyrosine and tyrosyl-alanine: solid-state IR-LD spectroscopy, quantum chemical calculations and vibrational analysis.
Koleva BB, Kolev TM, Spiteller M.
Spectrochimica Acta Part A: Molecular Spectroscopy, 68, 1187-1196 (2007)
Yufang Yang et al.
Frontiers in aging neuroscience, 10, 208-208 (2018-08-16)
Background: The deposition of β-sheet rich amyloid in senile plaques is a pathological hallmark of Alzheimer's disease (AD), which is thought to cause neuronal dysfunction. Previous studies have strongly implicated that intracerebral infusion of brain extract containing aggregated β-amyloid (Aβ)
Jenni Tienaho et al.
Molecules (Basel, Switzerland), 24(12) (2019-06-28)
Endophytes are microorganisms living inside plant hosts and are known to be beneficial for the host plant vitality. In this study, we isolated three endophytic fungus species from the roots of Scots pine seedlings growing on Finnish drained peatland setting.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service