Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5069

Sigma-Aldrich

4-Trifluoromethylumbelliferyl oleate

Synonym(s):

TFMU-oleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37F3O4
CAS Number:
Molecular Weight:
494.59
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

assay

≥98% (TLC)

form

liquid

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)Oc1ccc2c(OC(=O)C=C2C(F)(F)F)c1

InChI

1S/C28H37F3O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-26(32)34-22-18-19-23-24(28(29,30)31)21-27(33)35-25(23)20-22/h9-10,18-21H,2-8,11-17H2,1H3/b10-9-

InChI key

SUYMAHKATXKUBJ-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Fluorescent lipase substrate (ex. 385 nm, em. 502 nm [pH 8])

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service