Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T5000

Sigma-Aldrich

DL-β-(2-Thienyl)serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO3S
CAS Number:
Molecular Weight:
187.22
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

SMILES string

NC(C(O)c1cccs1)C(O)=O

InChI

1S/C7H9NO3S/c8-5(7(10)11)6(9)4-2-1-3-12-4/h1-3,5-6,9H,8H2,(H,10,11)

InChI key

GNUCLSFLDHLOBV-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Wapnir et al.
The Biochemical journal, 177(1), 347-352 (1979-01-01)
The inhibitory properties of beta-2-thienyl-dl-alanine on rat phenylalanine hydroxylase from crude liver and kidney homogenates were assessed in vitro and in vivo, as well as its effects on the intestinal transport of phenylalanine, by using a perfusion procedure in vivo.
P J Reis et al.
Australian journal of biological sciences, 38(2), 151-163 (1985-01-01)
Administration of the methionine analogue methoxinine (O-methyl-DL-homoserine) to sheep substantially changed the composition of wool; in addition wool fibres were weakened and the staple crimp frequency was reduced for a prolonged period. The proportions of high-tyrosine proteins were reduced by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service