T4914
Silicone rubber laboratory tubing
I.D. × O.D. 1/2 in. × 11/16 in.
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
I.D. × O.D.
1/2 in. × 11/16 in.
wall size
3/32 in.
General description
Translucent and flexible. Temp. range: -60 to 460°F (-51 to 238°C). Do not use with concentrated acids or dilute sodium hydroxide. Suitable for high-temperature applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)