Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T4779

Supelco

Tenocyclidine hydrochloride

Synonym(s):

1-(1-[2-Thienyl]cyclohexyl)piperidine, TCP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phencyclidine and psychotomimetic sigma opiates: recent insights into their biochemical and physiological sites of action.
M S Sonders et al.
Trends in neurosciences, 11(1), 37-40 (1988-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service