T4196
L-Tryptophan
Synonym(s):
(S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid
About This Item
Recommended Products
biological source
non-animal source
assay
≥98.5%
form
crystalline powder
manufacturer/tradename
Ajinomoto
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, heavy metals, residual solvents, tested
mp
280-285 °C (dec.) (lit.)
solubility
1 M HCl: 10 mg/mL
application(s)
pharmaceutical (small molecule)
foreign activity
cytotoxicity, tested
SMILES string
N[C@@H](Cc1c[nH]c2ccccc12)C(O)=O
InChI
1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1
Inchi Key
QIVBCDIJIAJPQS-VIFPVBQESA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Other Notes
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service