Key Documents
T3420
Eppendorf® Safe-Lock microcentrifuge tubes
volume 2.0 mL, yellow
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
cap (Safe-Lock lid)
polypropylene
sterility
non-sterile
feature
RCF 30,000 × g
packaging
case of 1000
pack of 500 ea
manufacturer/tradename
Eppendorf® 0030 120.205 (EU)
Eppendorf® 022363450 (USA)
volume
2.0 mL
color
yellow
Features and Benefits
Tubes snap closed and stay closed. Flat top with writing surface for positive sample identification
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)