Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3330

Sigma-Aldrich

Thermitase from Bacillus cereus

recombinant, expressed in Bacillus subtilis, enzyme ≥5.0 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

recombinant

expressed in Bacillus subtilis

form

solid

enzyme activity

≥5.0 units/mg protein

mol wt

33 kDa

foreign activity

DNase, RNase, none detected

storage temp.

−20°C

General description

Thermitase from Bacillus cereus is a protease that inactivates Rnases, Dnases and enzymes in reactions.

Application

Thermitase is used for the isolation of RNA and DNA (plasmid and genomic). Thermitase has been used in a study to assess the development and application of 15N-tracer substances for measuring the whole-body protein turnover rates. [1] Thermitase has also been used in a study to investigate a computational procedure for transferring a binding site onto an existing protein scaffold. [2]

Biochem/physiol Actions

Thermitase is a thermostable (4-60°C) serine protease that remains active over a wide pH range of 4-12.5. This protease retains its activity in the presence of EDTA, SDS, or Triton X-100.

Unit Definition

One unit is defined as the amount of enzyme required to solubilize 1 absorbance unit (595nm) of milk in 60 minutes at 37°C in a total reaction volume of 200ul.

Physical form

Supplied as a solution in 20mM Tris-HCl (pH 7.4), 1mM CaCl2 and 50% glycerol.

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

OptGraft: A computational procedure for transferring a binding site onto an existing protein scaffold
Fazelinia, H., et al.
Protein Science, 18, 180-195 (2008)
Klaus D Wutzke
Isotopes in environmental and health studies, 48(2), 239-258 (2012-03-09)
Our research group of the Children's Hospital of the University of Rostock (Rostock group) has long-time experience in (15)N-labelling and in using yeast protein and its hydrolysates for tracer kinetic studies to evaluate parameters of the whole-body protein metabolism in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service