Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3327

Sigma-Aldrich

Tubulysin A

≥95% (HPLC), from Streptomyces sp., film

Synonym(s):

TubA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H65N5O10S
CAS Number:
Molecular Weight:
844.07
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

biological source

Streptomyces sp.

Quality Level

assay

≥95% (HPLC)

form

film

color

colorless to white

solubility

organic solvents: soluble

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H]1CCCCN1C)C(=O)N(COC(=O)CC(C)C)[C@H](C[C@@H](OC(C)=O)c2nc(cs2)C(=O)N[C@H](C[C@H](C)C(O)=O)Cc3ccc(O)cc3)C(C)C

InChI

1S/C43H65N5O10S/c1-10-27(6)38(46-40(53)34-13-11-12-18-47(34)9)42(54)48(24-57-37(51)19-25(2)3)35(26(4)5)22-36(58-29(8)49)41-45-33(23-59-41)39(52)44-31(20-28(7)43(55)56)21-30-14-16-32(50)17-15-30/h14-17,23,25-28,31,34-36,38,50H,10-13,18-22,24H2,1-9H3,(H,44,52)(H,46,53)(H,55,56)/t27-,28-,31+,34+,35+,36+,38-/m0/s1

InChI key

IBEDDHUHZBDXGB-OEJISELMSA-N

Biochem/physiol Actions

Tubulysin A is a myxobacterial product that can function as an antiangiogenic agent in many in vitro assays1.
Tubulysin A is a novel antibiotic, which is anti-microtubule, anti-mitotic, apoptosis inducer, anticancer, anti-angiogenic, and antiproliferative.
Tubulysin A is a novel antibiotic, which is anti-microtubule, anti-mitotic, apoptosis inducer, anticancer, anti-angiogenic, and antiproliferative. Tubulysins are cytotoxic peptides, which include 9 members (A-I). Tubulysin A has potential application as an anticancer agent. It arrests cells in the G2/M phase. Tubulysin A inhibits polymerization more efficiently than vinblastine and induces depolymerization of isolated microtubules. Tubulysin A has potent cytostatic effects on various tumor cell lines with IC50 in the picomolar range.

Preparation Note

Tubulysin A is soluble in organic solvents.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurmeet Kaur et al.
The Biochemical journal, 396(2), 235-242 (2006-02-24)
Tubulysin A (tubA) is a natural product isolated from a strain of myxobacteria that has been shown to depolymerize microtubules and induce mitotic arrest. The potential of tubA as an anticancer and antiangiogenic agent is explored in the present study.
Lindsay M Steirer et al.
PloS one, 6(11), e27249-e27249 (2011-11-19)
Polysialic acid (PSA), an α2,8-linked homopolymer of N-acetylneuraminic acid (Neu5Ac), is developmentally regulated and its expression is thought to be restricted to a few tissues in adults. Recently, we showed that two human pathogens expressed a derivative of PSA containing
Christina M Van Itallie et al.
Molecular biology of the cell, 26(15), 2769-2787 (2015-06-13)
Assembly and sealing of the tight junction barrier are critically dependent on the perijunctional actin cytoskeleton, yet little is known about physical and functional links between barrier-forming proteins and actin. Here we identify a novel functional complex of the junction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service