Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3326

Sigma-Aldrich

Tamapin

recombinant, expressed in E. coli, ≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in E. coli

Quality Level

assay

≥95% (HPLC)

form

lyophilized powder

shipped in

wet ice

storage temp.

−20°C

Amino Acid Sequence

Ala-Phe-Cys-Asn-Leu-Arg-Arg-Cys-Glu-Leu-Ser-Cys-Arg-Ser-Leu-Gly-Leu-Leu-Gly-Lys-Cys-Ile-Gly-Glu-Glu-Cys-Lys-Cys-Val-Pro-Tyr

Biochem/physiol Actions

Member of the α-5.4 scorpion toxin family. Blocks KCa2 channels with selectivity for KCa2.2 versus KCa2.1 channels.
Tamapin strongly associates with Ca(2+)-activated K(+) (SK) channels and blocks SK-channel mediated currents in hippocampal neurons1.

Preparation Note

A stock solution of 1 μM can be prepared by adding 0.289 ml of any conventional buffer per μg of peptide.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Pedarzani et al.
The Journal of biological chemistry, 277(48), 46101-46109 (2002-09-20)
The biophysical properties of small conductance Ca(2+)-activated K(+) (SK) channels are well suited to underlie afterhyperpolarizations (AHPs) shaping the firing patterns of a conspicuous number of central and peripheral neurons. We have identified a new scorpion toxin (tamapin) that binds
Ricardo C Rodríguez de la Vega et al.
Toxicon : official journal of the International Society on Toxinology, 43(8), 865-875 (2004-06-23)
Much of our knowledge on K+-channels was elucidated using specific peptide ligands isolated from a number of venomous organisms. Recently, this field received a strong support and increased interest due to the solution of the three-dimensional structure of a couple

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service