T3301
Transforming Growth Factor-β1 Receptor II Soluble Fragment human
≥97% (SDS-PAGE), recombinant, expressed in mouse NSO cells, lyophilized powder
Synonym(s):
TGF-β1 RII
About This Item
recombinant
expressed in mouse NSO cells
assay
≥97% (SDS-PAGE)
form
lyophilized powder
impurities
endotoxin, tested
storage temp.
−20°C
General description
Recombinant, human TGF-β1 soluble receptor II will bind to TGF-β1, -β2, -β3, and -β5.
Application
Physical form
Analysis Note
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service