The antibody neutralizes receptor-ligand interaction. It shows no cross-reactivity with recombinant human Tie-1.
Immunogen
recombinant human Tie-2 extracellular domain expressed in NSO cells.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline, with 5% trehalose.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.