Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T311060

Sigma-Aldrich

DMT-dT-CPG column

pore size 500 Å, 50 nmol, configured for PerkinElmer, configured for ABI

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.51

Quality Level

type

for DNA synthesis

product line

Proligo Reagents

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

pore size

500 Å

capacity

≥99 % (Coupling Efficiency)

λ

conforms (UV/VIS Identity)

compatibility

configured for ABI
configured for PerkinElmer

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1)-c2ccccc2C3=NC(C)(C)CO3

InChI

1S/C18H19NO/c1-13-8-10-14(11-9-13)15-6-4-5-7-16(15)17-19-18(2,3)12-20-17/h4-11H,12H2,1-3H3

InChI key

JUNZDJNCZJUDRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service