Key Documents
T3045
Eppendorf® Safe-Lock microcentrifuge tubes
volume 2.0 mL, blue
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Product T3045 is not currently sold in your country. Contact Technical Service
material
cap (Safe-Lock lid)
polypropylene
sterility
non-sterile
feature
RCF 30,000 × g
packaging
case of 1,000
pack of 500
manufacturer/tradename
Eppendorf® 0030 120.221 (EU)
Eppendorf® 022363395 (USA)
volume
2.0 mL
color
blue
Features and Benefits
Tubes snap closed and stay closed. Flat top with writing surface for positive sample identification
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)