T3031
Microcentrifuge tubes, extended capacity
volume 2.0 mL , graduated, non-siliconized polypropylene
All Photos(1)
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
Recommended Products
material
cap
non-siliconized polypropylene
sterility
non-sterile
packaging
pack of 400
volume
2.0 mL , graduated
General description
The tubes listed below are specifically designed to hold more volume than other comparable tubes while still fitting all of the most commonly used microcentrifuges. Writing surface on flat lids and side wall for quick sample ID.
The lids provide an easily punctured syringe port for easy access to the contents without opening the tube. Siliconized tubes simplify research requiring low adhesion surface.
The lids provide an easily punctured syringe port for easy access to the contents without opening the tube. Siliconized tubes simplify research requiring low adhesion surface.
Choose from one of the most recent versions:
Certificates of Analysis (COA)