Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2942

Sigma-Aldrich

Monoclonal Anti-TRANCE antibody produced in mouse

clone 70525, purified immunoglobulin, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

70525, monoclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 8-25 μg/mL

isotype

IgG2b

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFSF11(8600)

General description

TRANCE (TNF-related activation-induced cytokine), also called RANKL (RANK ligand), OPGL (Osteoprotegrin ligand), and ODF (Osteoclast differentiation factor) is a member of tumor necrosis factor superfamily that is expressed on the surface of osteoblasts and is released by activated T cells. When TRANCE binds to its receptor RANK it stimulates the formation, activation, survival and adherence of osteoclasts that increases the bone resorption. These effects are inhibited when RANKL binds to OPG that leads to decrease in bone resorption process and increase in bone volume and density. The TRANCE (RANKL)/RANK/OPG collectively activates NF-κB, JNK, Akt, p38, ERK, NFATc1 pathways. This axis is vital for maintaining the bone density, dendritic cell survival, B cell differentiation, mammary gland development. This signaling pathway maintains the osteoclast biology and is a therapeutic target for osteoporosis. Monoclonal Anti-TRANCE antibody recognizes human TRANCE (~27 kDa). This antibody demonstrates 5% cross reactivity with recombinant mouse TRANCE.

Specificity

The antibody show <5% cross-reactivity with recombinant mouse TRANCE.

Immunogen

recombinant human TRANCE

Application

Anti-TRANCE antibody may be used for immunohistochemistry of paraffin-embedded tissue sections at a working concentration of 8-25 μg/ml. For ELISA a concentration of 0.5-1.0 μg/ml is recommended. The antibody is also suitable for neutralization reactions (ND50 is 0.02-0.06 μg/ml).

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William J Boyle et al.
Nature, 423(6937), 337-342 (2003-05-16)
Osteoclasts are specialized cells derived from the monocyte/macrophage haematopoietic lineage that develop and adhere to bone matrix, then secrete acid and lytic enzymes that degrade it in a specialized, extracellular compartment. Discovery of the RANK signalling pathway in the osteoclast
Daniel Castellano et al.
The oncologist, 16(2), 136-145 (2011-02-03)
The diagnosis of bone metastases is an event with certain consequences for the patient. They often mean pain and can also mean pathological fractures, hypercalcemia, and spinal cord compression, all synonymous with a diminished quality of life and often also
Joel Jules et al.
Expert opinion on therapeutic targets, 14(9), 923-934 (2010-08-04)
Osteoporosis has become a worldwide health and social issue due to an aging population. Four major antiresorptive drugs (agents capable of inhibiting osteoclast formation and/or function) are currently available on the market: estrogen, selective estrogen receptor modulators (SERMs), bisphosphonates and
David J Mellis et al.
The Journal of endocrinology, 211(2), 131-143 (2011-09-10)
Osteoclasts are the specialised cells that resorb bone matrix and are important both for the growth and shaping of bones throughout development as well as during the process of bone remodelling that occurs throughout life to maintain a healthy skeleton.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service