Skip to Content
MilliporeSigma

T2830

Tomatine hydrate

≥95% (HPLC)

Synonym(s):

Lycopersicin, NSC 234440, NSC 9223, Spirosolane, a-Tomatine, alpha-tomatine, b-D-galactopyranoside deriv

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C50H83NO21 · xH2O
CAS Number:
Molecular Weight:
1034.19 (anhydrous basis)
EC Number:
241-429-5
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

room temp

Application

Tomatine has been used as a standard for HPLC-based determination of tomatine from red and green tomatoes1. It has also been used to study its teratogenic effect using frog embryos2.

Biochem/physiol Actions

When the pure compounds were isolated, alpha-Tomatine was highly effective in inhibiting all of the cell lines, whereas dehydrotomatine, tomatidenol, and tomatidine had little, if any, effect on cell inhibition. It had been previously shown that commercial tomatine, which is customarily an approximate 10:1 mixture of α-tomatine and dehydrotomatine, strongly inhibits the growth of both colon and liver cancer cells.
It had been previously shown that commercial tomatine, which is customarily an approximate 10:1 mixture of α-tomatine and dehydrotomatine, strongly inhibits the growth of both colon and liver cancer cells.

Preparation Note

Tomatine hydrate is soluble in DMSO at a concentration that is greater than 20 mg/ml.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library