Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2827

Sigma-Aldrich

Anti-dTAF80 antibody, Mouse monoclonal

clone 3D10, purified from hybridoma cell culture

Synonym(s):

Anti-TAF5, Anti-TAF80/85, Anti-TAFII80, Anti-dmTAF5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

3D10, monoclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

Drosophila melanogaster

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

indirect ELISA: suitable
western blot: 8-16 μg/mL using nuclear extracts of D.Mel cells.

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Drosophila melanogaster ... Taf5(47900)

General description

Monoclonal Anti-dTAF80 (mouse IgG2b isotype) is derived from the hybridoma 3D10 produced by the fusion of mouse myeloma cells and splenocytes from mice. Drosophila tightly associated factor 80 (dTAF80), also known as tightly associated factor 5 (TAF5) consists of numerous copies ofβ−transducin (WD40) repeat.

Specificity

Monoclonal Anti-dTAF80 reacts specifically with Drosophila dTAF80

Immunogen

Drosophila TFIID complex.

Application

Anti-dTAF80 antibody, mouse monoclonal may be used in immunoblotting and enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Drosophila tightly associated factor 80 (dTAF80) recognizes and interacts with a higher-order complex containing TATA box-binding protein (TBP), delta N250, 110 and 60.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples shouldbe discard if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The dTAF II 80 subunit of Drosophila TFIID contains beta-transducin repeats
Dynlacht B D, et al.
Nature, 363(6425), 176-179 (1993)
Molecular cloning of a novel 120-kDa TBP-interacting protein
Yogosawa S, et al.
Biochemical and Biophysical Research Communications, 229(2), 612-617 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service