T2795
Eppendorf® Safe-Lock microcentrifuge tubes
volume 2.0 mL, natural
About This Item
Recommended Products
material
cap (Safe-Lock lid)
polypropylene
sterility
non-sterile
feature
RCF 30,000 × g
manufacturer/tradename
Eppendorf® 0030 120.094 (EU)
Eppendorf® 022363352 (USA)
volume
2.0 mL
color
natural
Features and Benefits
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)