Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2660

Sigma-Aldrich

Trizma® citrate tribasic solution

1 M

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H11NO3)3 . C6H8O7
CAS Number:
Molecular Weight:
555.53
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

form

liquid

Quality Level

concentration

1 M

SMILES string

NC(CO)(CO)CO.NC(CO)(CO)CO.NC(CO)(CO)CO.OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7.3C4H11NO3/c7-3(8)1-6(13,5(11)12)2-4(9)10;3*5-4(1-6,2-7)3-8/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);3*6-8H,1-3,5H2

InChI key

OHSJHRFFVBSWTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service