T2539
Silicone rubber laboratory tubing
I.D. × O.D. 1/8 in. × 3/8 in.
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
Recommended Products
I.D. × O.D.
1/8 in. × 3/8 in.
wall size
1/8 in.
General description
Translucent and flexible. Temp. range: -60 to 460°F (-51 to 238°C). Do not use with concentrated acids or dilute sodium hydroxide. Suitable for high-temperature applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)