Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2440

Sigma-Aldrich

Anti-Tumor Necrosis Factor Soluble Receptor II antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-TNF sRII

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

Tumor Necrosis Factor Receptor II (TNF RII) is a 75kD transmembrane glycoprotein expressed mainly on endothelial cells, macrophages, Langerhans cells and monocytes. It has a crucial role in regulating TNF α function by antagonizing its biological activity that is by blocking TNF-α-induced apoptosis. Anti-Tumor Necrosis Factor Soluble Receptor II (TNF sRII) antibody can be used in western blotting. Goat anti-Tumor Necrosis Factor Soluble Receptor II antibody reacts specifically with mouse TNF sRII. This antibody has also shown less than 15% cross-reactivity for recombinant human TNF-sRII.

Specificity

The antibody detects mouse TNF RII and shows <15% cross-reactivity with recombinant human TNF sRII.

Immunogen

recombinant mouse TNF soluble receptor II (TNF sRII) expressed in Escherichia coli.

Application

Anti-Tumor Necrosis Factor Soluble Receptor II (TNF sRII) antibody can be used in indirect ELISA.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Lien et al.
European journal of immunology, 25(9), 2714-2717 (1995-09-01)
Lipopolysaccharide (LPS), a part of the Gram-negative bacteria cell wall, is a potent inducer of tumor necrosis factor (TNF). TNF is an important mediator in Gram-negative infections such as meningococcal septic shock, but its harmful action can be prevented by
J R Bradley et al.
The American journal of pathology, 146(1), 27-32 (1995-01-01)
We have used confocal immunofluorescence microscopy and immunogold electron microscopy to localize tumor necrosis factor (TNF) receptors in cultured human umbilical vein endothelial cells. The 75-kd receptor is largely expressed on the cell surface and undergoes endocytosis and transport to
B Wang et al.
Immunology, 88(2), 284-288 (1996-06-01)
Langerhans' cells (LC) represent the major antigen-presenting cells within the epidermis. Following exposure of the skin to antigen, LC take up antigen, migrate into draining lymph nodes (DLN) and present processed antigen to T lymphocytes, thereby initiating an immune response.
Begoña Lainez et al.
International immunology, 16(1), 169-177 (2003-12-23)
Tumor necrosis factor (TNF)-alpha is a pleiotropic cytokine involved in a broad spectrum of inflammatory and immune responses including proliferation, differentiation and cell death induction in several cell types. The biological effects of TNF-alpha are mediated via the cell-surface TNF

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service