Key Documents
T2406
Sigma-Aldrich
Ultramicro pipette tips
Microcapillary, flat tip, maximum volume 10 μL, sterile
About This Item
UNSPSC Code:
41121606
material
colorless
colorless tip
polypropylene
sterility
sterile
feature
barrier: no
packaging
pack of 200 ea (Trays)
pack of 200 tips (10 trays of 20 tips)
manufacturer/tradename
Sigma-Aldrich
maximum volume
10 μL
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Application
for loading sequencing gels in vertical slab gels as thin as 0.4 mm.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)