Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T2190

Sigma-Aldrich

Anti-Tumor Necrosis Factor Soluble Receptor I antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-TNFsRI

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunohistochemistry (frozen sections): suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

The antibody recognizes cell surface mouse TNF RI (55 kDa) and exhibits TNF agonist activities. The antibody shows < 5% cross-reactivity with recombinant human TNF sRI and no cross-reactivity with recombinant human TNFsRII.

Immunogen

recombinant mouse TNF soluble receptor I, expressed in Escherichia coli.

Biochem/physiol Actions

The biological effects of tumor necrosis factor soluble factor (TNSF) are mediated by either type I or type II receptors. Type I receptors that are ubiquitously present on all cells whereas type II receptors are expressed only on cells of immune system and endothelial cells. Receptors of TNFSF such as TNFR1 and Fas have their own death domains that activate the apoptosis. Other receptors such as TNFR2 do not have death domains and utilize TNF associated factors (TRAFs) to activate intracellular signaling. The pathways activated via TNFsRI include NF-κB, JNK and MAPK.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Lotz et al.
Journal of leukocyte biology, 60(1), 1-7 (1996-07-01)
Receptors in the nerve growth factor/tumor necrosis factor receptor family are characterized by the presence of cysteine-rich motifs of approximately 40 amino acids in the extracellular domain. The ligands are type II transmembrane proteins with beta-strands that form a jelly-roll
B B Aggarwal
Annals of the rheumatic diseases, 59 Suppl 1, i6-16 (2000-10-29)
Tumour necrosis factor (TNF) is a pleiotropic cytokine that mediates apoptosis, cell proliferation, immunomodulation, inflammation, viral replication, allergy, arthritis, septic shock, insulin resistance, autoimmune diseases, and other pathological conditions. TNF transduces these cellular responses through two distinct receptors: type I

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service