Key Documents
T2166
Sigma-Aldrich
Quartz digital timers
Model T-440
About This Item
UNSPSC Code:
41122411
NACRES:
NB.35
size
2 1/2 in. × 2 3/4 in. × 3/4 in.
battery power
1.5 V button cell
General description
Display: 6 digits, 0.75 inch
Special features: four independent channels each offer 100 hour countdown and countup with buzzer alarm; memory recall function; clock, magnet, clip and stand on back of unit
Choose from one of the most recent versions:
Certificates of Analysis (COA)