T2156
Ultramicro pipette tips
Microcapillary, round tip, maximum volume 200 μL, sterile
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
Recommended Products
material
colorless
polypropylene
sterility
sterile
feature
barrier: no
packaging
pack of 200 ea (Trays)
manufacturer/tradename
Sigma-Aldrich
maximum volume
200 μL
General description
Low capacity improves accuracy with small sample volumes by reducing dead space.
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)