T1654
Transforming Growth Factor-β1 from human platelets
TGF-β1, lyophilized powder, suitable for cell culture, 1 × 106 U/mg
Synonym(s):
TGF-β1
About This Item
Recommended Products
biological source
human
assay
≥97% (SDS-PAGE and N-terminal analysis)
form
lyophilized powder
potency
0.04-0.3 ng/mL ED50/EC50
quality
endotoxin tested
specific activity
1 × 106 U/mg
mol wt
protein 25 kDa
packaging
pkg of 1 μg
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
technique(s)
cell culture | mammalian: suitable
impurities
≤1 EU/μgtested (LAL test)
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... TGFB1(7040)
General description
Application
- to study the induction of a-smooth muscle actin by transforming growth factor-1 (TGF-β1) in breast cancer cells.[4]
- to study the efficiency of TGF-β1 in promoting embryonic stem-like cell proliferation with or without feeders.[5]
- as control to analyse the isolated TGF-β1 HPLC using reverse-phase chromatography.[6]
- to induce profibrotic response in renal fibroblasts and further study functional role for Smad3 (mothers against decapentaplegic homolog 3) T388 phosphorylation.[7]
Biochem/physiol Actions
Physical form
Analysis Note
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service