Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T111N0-HH

Sigma-Aldrich

NPPOC-dT RayDite

Synonym(s):

NPPOC-dT Amidite, NPPOC-dT Phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H40N5O10P
CAS Number:
Molecular Weight:
649.63
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

type

for DNA synthesis

product line

Proligo Reagents

assay

≥96% (31P-NMR)
≥96.0% (reversed phase HPLC)

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for NMR (13C-NMR)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)O[C@H]1C[C@@H](O[C@@H]1COC(=O)OCC(C)c2ccccc2[N+]([O-])=O)N3C=C(C)C(=O)NC3=O

InChI

1S/C29H40N5O10P/c1-18(2)33(19(3)4)45(42-13-9-12-30)44-24-14-26(32-15-20(5)27(35)31-28(32)36)43-25(24)17-41-29(37)40-16-21(6)22-10-7-8-11-23(22)34(38)39/h7-8,10-11,15,18-19,21,24-26H,9,13-14,16-17H2,1-6H3,(H,31,35,36)/t21?,24-,25+,26+,45?/m0/s1

InChI key

ZOIPTUUCAAKGFT-GPZIPZSXSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

This product and it use are covered under US Patent Number 5,763,599
RayDite is a trademark of Sigma-Aldrich Co. LLC

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service