Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0534

Sigma-Aldrich

anti-T2-toxin

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

conjugate

unconjugated

antibody form

whole antiserum

storage temp.

−20°C

target post-translational modification

unmodified

Diagnostic Implications

Mycotoxins, a group of chemically diverse secondary fungal metabolites, induce a variety of toxic responses in humans and animals when food and feed containing these compounds are ingested. Rapidly quantitating levels of mycotoxin contamination can help reduce exposure to these toxins. Polyclonal antibodies are developed using KLH or BSA conjugates. The antiserum is fractionated (containing primarily the gamma globulin (iG) fraction of antiserum) and supplied liquid in 0.01 M phosphate buffered saline, pH 7.4. Peroxidase conjugates of the antibody are produced using fractionated antisera. Monoclonal antibodies are supplied as delipidized, filtered ascites fluid.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service