Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0387

Sigma-Aldrich

L-Threonine–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817

form

saline suspension

extent of labeling

2-10 μmol per mL

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Application

Threonine-agarose is used for protein chromatography, affinity chromatography and amino acid resins. Threonine-agarose has been used for the purification and characterization of a soluble phosphatidylinositol 4-kinase from the yeast Saccharomyces cerevisiae.

Physical form

Suspension in 2.0 M NaCl containing preservative

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C A Flanagan et al.
The Journal of biological chemistry, 267(33), 24117-24125 (1992-11-25)
A phosphatidylinositol (PI) 4-kinase was purified 25,000-fold from the cytosolic fraction of extracts from the yeast Saccharomyces cerevisiae. The purification consisted of an ammonium sulfate fractionation followed by chromatography on sulfonated-agarose (S-Sepharose), phosphocellulose, threonine-agarose, and quaternary amino (Mono Q), and
Lylla Ngo et al.
Acta histochemica, 105(1), 21-28 (2003-04-02)
This study describes a novel rapid histochemical method to survey surface properties of sperm in a model system, the sea urchin. Surface properties of live and fixed sea urchin sperm of 2 species (Strongylocentrotus purpuratus and Lytechinus pictus) in seawater

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service