Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T0206

Sigma-Aldrich

Tyrosinase Human, Recombinant

Synonym(s):

Tyrosinase Human, Recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

assay

≥95% (reversed phase HPLC)

form

buffered aqueous solution

specific activity

5 U/μL

mol wt

60 kDa

NCBI accession no.

storage temp.

2-8°C

Application

Study enzyme kinetics and screen for inhibitors.

Biochem/physiol Actions

Tyrosinase is a copper-containing enzyme that catalyzes four steps in the melanin and catecholamine synthesis pathways. It catalyzes the formation of L-DOPA (L-3,4-dihydroxyphenylalanine) in the brain and nervous system. It oxidizes monophenols and o-diphenols to o-quinones. The molecular mass is approximately 60 kDa.

Unit Definition

One unit will produce 100 picomoles of dopachrome-MBTH adduct from L-tyrosine per minute at pH 7.3 at 37 °C.

Physical form

Supplied in 30mM NaPO4, pH 6.8

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Murtas et al.
Histochemistry and cell biology, 151(2), 175-185 (2018-09-21)
While histological analysis represents a powerful tool for the classification of melanocytic lesions as benign or malignant, a clear-cut distinction between a nevus and a melanoma is sometimes a challenging step of the diagnostic process. The immunohistochemical detection of tyrosinase
Atsushi Ishihara et al.
Bioscience, biotechnology, and biochemistry, 82(1), 22-30 (2018-01-04)
Tyrosinase is the key enzyme that controls melanin formation in the human skin. We performed a screening of 96 extracts of mushroom cultures and fruiting bodies for examining their inhibitory activity against mushroom tyrosinase. The ethyl acetate extracts of culture
Michele Schlich et al.
Colloids and surfaces. B, Biointerfaces, 171, 675-681 (2018-08-15)
Tyrosinase is one of the key enzymes in mammalian melanin biosynthesis. Decreasing tyrosinase activity has been targeted for the prevention of conditions related to the hyperpigmentation of the skin, such as melasma and age spots. This paper is devoted to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service