Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP8041

Sigma-Aldrich

LAG-3 (mouse): FC (mouse)

recombinant, expressed in CHO cells, >95% (SDS-PAGE)

Synonym(s):

ACEH, FDC protein, Lymphocyte activation gene-3

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$571.00

$571.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
$571.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$571.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

mouse

recombinant

expressed in CHO cells

assay

>95% (SDS-PAGE)

form

liquid

mol wt

monomer ~180 kDa by SDS-PAGE (non-reducing)
monomer ~80 kDa by SDS-PAGE (reducing)

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

≥0.2 mg/mL

impurities

<1 EU/μg endotoxin, tested

color

clear

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Lag3(16768)

General description

Lymphocyte activation gene 3 (LAG-3; CD223) plays an important role in negatively regulating T cell proliferation, function and homeostasis. It is required for maximal natural and induced regulatory T cell (Treg) function. LAG-3 is closely related to the T cell co-receptor CD4 and binds to MHC class II molecules but with a significantly higher affinity than CD4.

Physical form

Solution in PBS.

Other Notes

The sequence coding for the 4 extracellular Ig-like domains of mouse LAG-3 (D1-D4) is fused to the Fc portion of mouse IgG2a (hinge-CH2-CH3).

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service