Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP8031

Sigma-Aldrich

FNDC5

recombinant, expressed in E. coli, untagged, >95% (SDS-PAGE)

Synonym(s):

Fibronectin type III domain-containing protein 5, Fibronectin type III repeat-containing protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

conjugate

untagged

assay

>95% (SDS-PAGE)

form

lyophilized

mol wt

~16 kDa

packaging

pkg of 10 μg

impurities

<0.01 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FNDC5(252995)

General description

Irisin is a recently described exercise-induced hormone secreted by skeletal muscle in mice and humans. Irisin activates beige fat cells (beige cells have a gene expression pattern distinct from either white or brown fat and are preferentially sensitive to the polypeptide hormone irisin). Irisin is cleaved from the type I membrane protein FNDC5 and improves systemic metabolism by increasing energy expenditure.

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 100 μL deionized water.

Other Notes

FNDC5 (extracellular domain, human (aa 32-152) / mouse (aa 29-149)) is untagged. FNDC5 extracellular domain has 100% identity between human, mouse and rat.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service